Product Details

SNP ID
rs6956899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151471448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCAACATAATAGGTATTCTATTT[A/G]TAAGAAAAAAAAGACAAACCAGTAA
Phenotype
MIM: 601293
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHEB PubMed Links

Gene Details

Gene
RHEB
Gene Name
Ras homolog enriched in brain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005614.3 Intron NP_005605.1
XM_011516457.1 Intron XP_011514759.1

View Full Product Details