Product Details

SNP ID
rs11831564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:80717935 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAATGAGGAAATGGAAAGAAATAC[C/G]CACACGGCCCCAGTTCCTGCTCCAA
Phenotype
MIM: 159990 MIM: 159991
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYF5 PubMed Links

Gene Details

Gene
MYF5
Gene Name
myogenic factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005593.2 Intron NP_005584.2
Gene
MYF6
Gene Name
myogenic factor 6
There are no transcripts associated with this gene.

View Full Product Details