Product Details

SNP ID
rs521476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33244758 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCCTCTGTTACACAGCAGTAAGA[G/T]AGGGGTGGAGAGGAGTTAAAATCAT
Phenotype
MIM: 601497 MIM: 613929
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BAG1 PubMed Links

Gene Details

Gene
BAG1
Gene Name
BCL2 associated athanogene 1
There are no transcripts associated with this gene.

Gene
SPINK4
Gene Name
serine peptidase inhibitor, Kazal type 4
There are no transcripts associated with this gene.

View Full Product Details