Product Details

SNP ID
rs12747477
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:54982575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGACCCACACAGAAAATGACCCC[A/G]ACCCCATGTGAGAACACGCAAAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM61 PubMed Links

Gene Details

Gene
TMEM61
Gene Name
transmembrane protein 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182532.2 Intron NP_872338.1
XM_005270586.4 Intron XP_005270643.1
XM_011540911.2 Intron XP_011539213.1
XM_011540912.2 Intron XP_011539214.1

View Full Product Details