Product Details

SNP ID
rs9394767
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:41192167 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGGGGTCTGCAGCTCCGAGCAGAA[A/G]CCACTGGTCACATTTCCTCGGGACT
Phenotype
MIM: 609715
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TREML2 PubMed Links

Gene Details

Gene
TREML2
Gene Name
triggering receptor expressed on myeloid cells like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024807.3 1130 UTR 3 NP_079083.2
XM_011514917.2 1130 UTR 3 XP_011513219.1

View Full Product Details