Product Details

SNP ID
rs3764942
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:136133811 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCATGTGGCGCGCCCGTCTTAAGT[T/C]TGAGCATGCTCAGTAGCCAAAACAG
Phenotype
MIM: 603110
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SMAD5 PubMed Links

Gene Details

Gene
SMAD5
Gene Name
SMAD family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001419.2 Intron NP_001001419.1
NM_001001420.2 Intron NP_001001420.1
NM_005903.6 Intron NP_005894.3
XM_017009470.1 Intron XP_016864959.1
Gene
SMAD5-AS1
Gene Name
SMAD5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details