Product Details

SNP ID
rs10459237
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:50848352 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTCCCCACTTCCCACCCAGAAATC[A/G]GTTTAGTCTCTAAGAACCAGTCATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS12 PubMed Links

Gene Details

Gene
TMPRSS12
Gene Name
transmembrane protease, serine 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182559.2 Intron NP_872365.1

View Full Product Details