Product Details

SNP ID
rs9749384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11723906 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTGCAAAGACTGAATAGCTATG[C/T]CACATTTAACTATATGTTCCTCGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF823 PubMed Links

Gene Details

Gene
ZNF823
Gene Name
zinc finger protein 823
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080493.3 Intron NP_001073962.1
NM_001297610.1 Intron NP_001284539.1
NM_017507.1 Intron NP_059977.1
XM_017026941.1 Intron XP_016882430.1
XM_017026942.1 Intron XP_016882431.1

View Full Product Details