Product Details

SNP ID
rs3764510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74499611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTAAAAATTTTTATTAAAGTACC[A/G]GTAAAACTAAAAGTGCCTACTTTTA
Phenotype
MIM: 169800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNDP2 PubMed Links

Gene Details

Gene
CNDP2
Gene Name
CNDP dipeptidase 2 (metallopeptidase M20 family)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168499.1 24 UTR 5 NP_001161971.1
NM_018235.2 24 Intron NP_060705.2
XM_005266728.2 24 Intron XP_005266785.1
XM_006722503.2 24 Intron XP_006722566.1
XM_011526071.2 24 Intron XP_011524373.1
XM_011526072.2 24 Intron XP_011524374.1
XM_011526073.2 24 Intron XP_011524375.1

View Full Product Details