Product Details

SNP ID
rs514438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43328245 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGCCACTGTCTTCAGCTGTTTTC[C/T]GTAGTGATTGTTTCTAGGTATTTTA
Phenotype
MIM: 611246
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAL PubMed Links

Gene Details

Gene
ADAL
Gene Name
adenosine deaminase like
There are no transcripts associated with this gene.

Gene
LCMT2
Gene Name
leucine carboxyl methyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014793.4 2378 UTR 3 NP_055608.2

View Full Product Details