Product Details

SNP ID
rs456520
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:9917083 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACAGAGAGGGGCGGGAACAGCCAGC[A/T]GCCTGCCGGGAGCCAAAACGAAAGC
Phenotype
MIM: 610925 MIM: 614995
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IL17RC PubMed Links

Gene Details

Gene
IL17RC
Gene Name
interleukin 17 receptor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203263.1 10 UTR 5 NP_001190192.1
NM_001203264.1 10 UTR 5 NP_001190193.1
NM_001203265.1 10 UTR 5 NP_001190194.1
NM_032732.5 10 UTR 5 NP_116121.2
NM_153460.3 10 UTR 5 NP_703190.1
NM_153461.3 10 Intron NP_703191.1
XM_017007326.1 10 Intron XP_016862815.1
XM_017007327.1 10 Intron XP_016862816.1
XM_017007328.1 10 Intron XP_016862817.1
XM_017007329.1 10 Intron XP_016862818.1
XM_017007330.1 10 Intron XP_016862819.1
XM_017007331.1 10 Intron XP_016862820.1
XM_017007332.1 10 Intron XP_016862821.1
XM_017007333.1 10 Intron XP_016862822.1
XM_017007334.1 10 Intron XP_016862823.1
XM_017007335.1 10 Intron XP_016862824.1
XM_017007336.1 10 Intron XP_016862825.1
XM_017007337.1 10 Intron XP_016862826.1
XM_017007338.1 10 Intron XP_016862827.1
XM_017007339.1 10 Intron XP_016862828.1
XM_017007340.1 10 Intron XP_016862829.1
XM_017007341.1 10 Intron XP_016862830.1
XM_017007342.1 10 Intron XP_016862831.1
XM_017007343.1 10 Intron XP_016862832.1
XM_017007344.1 10 Intron XP_016862833.1
XM_017007345.1 10 Intron XP_016862834.1
XM_017007346.1 10 Intron XP_016862835.1
XM_017007347.1 10 Intron XP_016862836.1
XM_017007348.1 10 Intron XP_016862837.1
XM_017007349.1 10 Intron XP_016862838.1
Gene
IL17RE
Gene Name
interleukin 17 receptor E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193380.1 10 Intron NP_001180309.1
NM_153480.1 10 Intron NP_705613.1
NM_153481.1 10 Intron NP_705614.1
NM_153483.2 10 Intron NP_705616.2
XM_006712975.1 10 Intron XP_006713038.1
XM_006712976.1 10 Intron XP_006713039.1
XM_011533361.2 10 Intron XP_011531663.1
XM_011533362.2 10 Intron XP_011531664.1
XM_011533363.2 10 Intron XP_011531665.1
XM_011533364.2 10 Intron XP_011531666.1

View Full Product Details