Product Details

SNP ID
rs1048535
Assay Type
Functionally Tested
NCBI dbSNP Submissions
60
Location
Chr.1:152804989 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCTGAGTTTCTGGACTCCAGGGA[A/G]AAGATATGCTCTTGGCAAGTTCAGA
Phenotype
MIM: 612604 MIM: 612605 MIM: 612606
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LCE1B PubMed Links

Gene Details

Gene
LCE1B
Gene Name
late cornified envelope 1B
There are no transcripts associated with this gene.

Gene
LCE1C
Gene Name
late cornified envelope 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276331.1 451 UTR 3 NP_001263260.1
NM_178351.3 451 UTR 3 NP_848128.1
Gene
LCE1D
Gene Name
late cornified envelope 1D
There are no transcripts associated with this gene.

View Full Product Details