Product Details
- SNP ID
-
rs1509599
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:24501685 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCATCCCTGTAGACATGTCTTTGA[C/T]GTTTCATTATCATGCAGTAAGAATG
- Phenotype
-
MIM: 608178
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LUZP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78961907] are located under a probe and SNP(s) [rs78573266] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LUZP2
- Gene Name
- leucine zipper protein 2
View Full Product Details