Product Details

SNP ID
rs13296417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101539009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGACAGAAGAGAACAGGATCCT[G/T]AGCACAAGCTGAGGTTTTAAGAATG
Phenotype
MIM: 607699
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RNF20 PubMed Links

Gene Details

Gene
RNF20
Gene Name
ring finger protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019592.6 Intron NP_062538.5
XM_011518862.1 Intron XP_011517164.1

View Full Product Details