Product Details

SNP ID
rs3888651
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:42657628 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTGGATTTCTTCACAGGTTTCAAC[G/T]TCTGCAGCAGCCACATGGCTGTTGC
Phenotype
MIM: 161565
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928323 PubMed Links

Gene Details

Gene
LOC101928323
Gene Name
uncharacterized LOC101928323
There are no transcripts associated with this gene.

Gene
NKTR
Gene Name
natural killer cell triggering receptor
There are no transcripts associated with this gene.

Gene
ZBTB47
Gene Name
zinc finger and BTB domain containing 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145166.3 Intron NP_660149.2

View Full Product Details