Product Details

SNP ID
hCV3098774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8066200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGATGCGCTCGGCCCGGCCCA[G/T]GTGTGAGAGGTTCAGGCCCAAGGTC
Phenotype
MIM: 602565 MIM: 608529
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCL25 PubMed Links

Gene Details

Gene
CCL25
Gene Name
C-C motif chemokine ligand 25
There are no transcripts associated with this gene.

Gene
FBN3
Gene Name
fibrillin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321431.1 8289 Missense Mutation ATG,CTG M2717L NP_001308360.1
NM_032447.4 8289 Missense Mutation ATG,CTG M2717L NP_115823.3

View Full Product Details