Product Details

SNP ID
rs4972805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176147842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCGGGAGGTGGAGGCGCTAAAT[C/T]GAGGTCCCGGAAAGCTTTTCTGGGG
Phenotype
MIM: 142980 MIM: 142981 MIM: 611576
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXD3 PubMed Links

Gene Details

Gene
HOXD3
Gene Name
homeobox D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006898.4 Intron NP_008829.3
XM_005246509.3 Intron XP_005246566.1
XM_005246510.4 Intron XP_005246567.1
XM_005246511.3 Intron XP_005246568.1
XM_005246513.4 Intron XP_005246570.1
XM_006712477.2 Intron XP_006712540.1
XM_011511065.2 Intron XP_011509367.1
XM_011511066.2 Intron XP_011509368.1
Gene
HOXD4
Gene Name
homeobox D4
There are no transcripts associated with this gene.

Gene
MIR10B
Gene Name
microRNA 10b
There are no transcripts associated with this gene.

View Full Product Details