Product Details

SNP ID
rs5508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94563515 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATCGACTACCTGCTCCTCCTGCT[G/T]GTTGGACTACTGGCCCTTTCTCATG
Phenotype
MIM: 147935
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SERPINA4 PubMed Links

Gene Details

Gene
SERPINA4
Gene Name
serpin family A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289032.1 125 Silent Mutation CTG,CTT L48L NP_001275961.1
NM_001289033.1 125 Silent Mutation CTG,CTT L11L NP_001275962.1
NM_006215.3 125 Silent Mutation CTG,CTT L11L NP_006206.2

View Full Product Details