Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001282902.2 | 8620 | UTR 3 | NP_001269831.1 | ||
NM_001282903.2 | 8620 | UTR 3 | NP_001269832.1 | ||
NM_014112.4 | 8620 | UTR 3 | NP_054831.2 | ||
XM_005251049.2 | 8620 | UTR 3 | XP_005251106.1 | ||
XM_006716625.1 | 8620 | UTR 3 | XP_006716688.1 | ||
XM_011517264.1 | 8620 | UTR 3 | XP_011515566.1 | ||
XM_011517266.2 | 8620 | UTR 3 | XP_011515568.1 | ||
XM_011517267.1 | 8620 | UTR 3 | XP_011515569.1 | ||
XM_011517268.1 | 8620 | UTR 3 | XP_011515570.1 | ||
XM_017013801.1 | 8620 | UTR 3 | XP_016869290.1 |