Product Details

SNP ID
rs2280491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9772304 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAATAATAATAGCAATACCTATCT[C/G]TTAGGGAGGCACGAAATGATTAAGG
Phenotype
MIM: 616161
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DHRS7C PubMed Links

Gene Details

Gene
DHRS7C
Gene Name
dehydrogenase/reductase 7C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105571.2 Intron NP_001099041.1
NM_001220493.1 Intron NP_001207422.1

View Full Product Details