Product Details

SNP ID
rs6896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73375688 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTAAAGTACACAGTTTTAACA[C/T]ACTATCCACTCTATCAAGCAGACTT
Phenotype
MIM: 186360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANXA7 PubMed Links

Gene Details

Gene
ANXA7
Gene Name
annexin A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001156.4 1949 UTR 3 NP_001147.1
NM_001320879.1 1949 UTR 3 NP_001307808.1
NM_001320880.1 1949 UTR 3 NP_001307809.1
NM_004034.3 1949 UTR 3 NP_004025.1
XM_017016162.1 1949 UTR 3 XP_016871651.1
XM_017016163.1 1949 UTR 3 XP_016871652.1

View Full Product Details