Product Details

SNP ID
rs1141746
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:27666358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAAGAGTTAGGCCAAGAAGGAA[A/G]AAGAGGTTCTGGGAGCTGCTGGCTA
Phenotype
MIM: 147572
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFI6 PubMed Links

Gene Details

Gene
IFI6
Gene Name
interferon alpha inducible protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002038.3 551 UTR 3 NP_002029.3
NM_022872.2 551 UTR 3 NP_075010.1
NM_022873.2 551 UTR 3 NP_075011.1

View Full Product Details