Product Details

SNP ID
rs1379281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17078976 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATCTCCAGGGCCGTTTCTCACCC[A/G]CATCACCATTGACAGTTGGGGCTGT
Phenotype
MIM: 613434 MIM: 602129
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAUS8 PubMed Links

Gene Details

Gene
HAUS8
Gene Name
HAUS augmin like complex subunit 8
There are no transcripts associated with this gene.

Gene
MYO9B
Gene Name
myosin IXB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130065.1 Intron NP_001123537.1
NM_004145.3 Intron NP_004136.2

View Full Product Details