Product Details
- SNP ID
-
rs224395
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:35521599 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTTAGATTTAACGGGAAGTCCTT[A/G]GGAAGACAGATTTTTTTTTTTTTTT
- Phenotype
-
MIM: 609689
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C20orf173
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141862604] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C20orf173
- Gene Name
- chromosome 20 open reading frame 173
- Gene
- CEP250
- Gene Name
- centrosomal protein 250
There are no transcripts associated with this gene.
View Full Product Details