Product Details
- SNP ID
-
rs1184986
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:35618756 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCGGACCGAGCTGCCCGCCTCCCG[G/T]CCGCCCGAAGACCGACTCCCACGGC
- Phenotype
-
MIM: 612206
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FJX1
PubMed Links
Gene Details
- Gene
- FJX1
- Gene Name
- four jointed box 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014344.3 |
570 |
Silent Mutation |
CGG,CGT |
R40R |
NP_055159.2 |
View Full Product Details