Product Details

SNP ID
rs1046137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:61975026 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACTGATCAGCGTTCAGTTCACAC[A/G]CGCTCGAATTAAAGTCTAATGTGAG
Phenotype
MIM: 601796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAF4 PubMed Links

Gene Details

Gene
TAF4
Gene Name
TATA-box binding protein associated factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003185.3 4400 UTR 3 NP_003176.2

View Full Product Details