Product Details

SNP ID
rs1006050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:37694261 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCCTGTGGGACAGGTGCAGCAGC[A/G]AGTGGGAGGGGCTTTTGGAGCAGCT
Phenotype
MIM: 609045
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR141 PubMed Links

Gene Details

Gene
GPR141
Gene Name
G protein-coupled receptor 141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011515374.2 Intron XP_011513676.1
XM_011515375.2 Intron XP_011513677.1
XM_011515383.2 Intron XP_011513685.1
XM_011515385.2 Intron XP_011513687.1
XM_011515386.2 Intron XP_011513688.1
XM_011515388.2 Intron XP_011513690.1
XM_011515389.2 Intron XP_011513691.1
XM_017012160.1 Intron XP_016867649.1
XM_017012161.1 Intron XP_016867650.1
XM_017012162.1 Intron XP_016867651.1
XM_017012163.1 Intron XP_016867652.1
XM_017012164.1 Intron XP_016867653.1
XM_017012165.1 Intron XP_016867654.1
XM_017012166.1 Intron XP_016867655.1
XM_017012167.1 Intron XP_016867656.1

View Full Product Details