Product Details
- SNP ID
-
rs1015734
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:42396588 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATTCTCATTTTGTGTAGGTTCCA[A/G]AATTTGCTCATTAGGTTTGTGTTGC
- Phenotype
-
MIM: 605417
MIM: 610029
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DKK4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77829339] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DKK4
- Gene Name
- dickkopf WNT signaling pathway inhibitor 4
There are no transcripts associated with this gene.
- Gene
- VDAC3
- Gene Name
- voltage dependent anion channel 3
View Full Product Details