Product Details

SNP ID
rs12992405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:31174466 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGAAGGATGGCTAGCTTTTACAA[A/C]TCTGATGTAATCTTTACTTCCTCCC
Phenotype
MIM: 610229
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAPN14 PubMed Links

Gene Details

Gene
CAPN14
Gene Name
calpain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145122.1 2514 UTR 3 NP_001138594.1
NM_001321270.1 2514 UTR 3 NP_001308199.1
XM_011532863.2 2514 UTR 3 XP_011531165.1
XM_011532864.2 2514 UTR 3 XP_011531166.1
XM_011532865.1 2514 UTR 3 XP_011531167.1
XM_011532866.2 2514 UTR 3 XP_011531168.1

View Full Product Details