Product Details

SNP ID
rs1106201
Assay Type
Functionally tested
NCBI dbSNP Submissions
72
Location
Chr.1:205303798 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAAGGTGGTGGGGGCCGCGAGCTC[C/T]AAGGCTGTCTGGGAGAACTTGCCAT
Phenotype
MIM: 608131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUAK2 PubMed Links

Gene Details

Gene
NUAK2
Gene Name
NUAK family kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030952.2 959 Silent Mutation NP_112214.2
XM_005245515.4 959 Missense Mutation XP_005245572.1

View Full Product Details