Product Details

SNP ID
rs907603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1828103 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTCGACCTGGGATATGCATCTGC[A/G]TTCGAGGCAGGCAGTGGGGGTTAGG
Phenotype
MIM: 607719
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYT8 PubMed Links

Gene Details

Gene
SYT8
Gene Name
synaptotagmin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290332.1 Intron NP_001277261.1
NM_001290333.1 Intron NP_001277262.1
NM_001290334.1 Intron NP_001277263.1
NM_138567.4 Intron NP_612634.3
XM_011520455.1 Intron XP_011518757.1
XM_011520456.2 Intron XP_011518758.1
XM_011520457.2 Intron XP_011518759.1
XM_017018528.1 Intron XP_016874017.1
XM_017018529.1 Intron XP_016874018.1

View Full Product Details