Product Details

SNP ID
rs30454
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:39353536 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGGCATTATATTGTACATATTACT[A/G]TGTAGCTTATGGACACCCCTCCAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAMD4B PubMed Links

Gene Details

Gene
SAMD4B
Gene Name
sterile alpha motif domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303614.1 Intron NP_001290543.1
NM_018028.3 Intron NP_060498.2
XM_011527057.2 Intron XP_011525359.1
XM_011527060.2 Intron XP_011525362.1
XM_011527061.2 Intron XP_011525363.1
XM_011527062.2 Intron XP_011525364.1
XM_011527063.2 Intron XP_011525365.1
XM_011527064.2 Intron XP_011525366.1
XM_017026918.1 Intron XP_016882407.1
XM_017026919.1 Intron XP_016882408.1
XM_017026920.1 Intron XP_016882409.1
XM_017026921.1 Intron XP_016882410.1
XM_017026922.1 Intron XP_016882411.1
XM_017026923.1 Intron XP_016882412.1

View Full Product Details