Product Details

SNP ID
rs347519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43769364 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCTGACCTGGACAGGCATGGACA[C/T]GCACACACACAGCCGTGCAGAGAGG
Phenotype
MIM: 602754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNN4 PubMed Links

Gene Details

Gene
KCNN4
Gene Name
potassium calcium-activated channel subfamily N member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002250.2 Intron NP_002241.1
XM_005258882.2 Intron XP_005258939.1
XM_005258883.2 Intron XP_005258940.1
XM_011526938.1 Intron XP_011525240.1

View Full Product Details