Product Details

SNP ID
rs1047699
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10847109 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGATGTCTTGCTGGTCTGTCCCTC[C/T]GGAAGAATGATGATGCTTCCTGCAG
Phenotype
MIM: 605359
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PRH1-PRR4 PubMed Links

Gene Details

Gene
PRH1-PRR4
Gene Name
PRH1-PRR4 readthrough
There are no transcripts associated with this gene.

Gene
PRR4
Gene Name
proline rich 4 (lacrimal)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098538.2 398 Missense Mutation AGA,GGA R43G NP_001092008.2
NM_007244.2 398 Missense Mutation CAG,CGG Q120R NP_009175.2

View Full Product Details