Product Details

SNP ID
rs1045734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4270114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACGCACTCATGCCTGGCTCTCA[A/C]TTCCACTCCCGTAGAGCTAGCCCTA
Phenotype
MIM: 607927 MIM: 601569
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKFY1 PubMed Links

Gene Details

Gene
ANKFY1
Gene Name
ankyrin repeat and FYVE domain containing 1
There are no transcripts associated with this gene.

Gene
UBE2G1
Gene Name
ubiquitin conjugating enzyme E2 G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003342.4 3311 UTR 3 NP_003333.1

View Full Product Details