Product Details
- SNP ID
-
rs1802948
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:88899117 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGAAACCACACGCCCTACTTTGC[A/C]CTTTCCTGTCCATCCCAGACCTACT
- Phenotype
-
MIM: 602281
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HAPLN3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113688804] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HAPLN3
- Gene Name
- hyaluronan and proteoglycan link protein 3
There are no transcripts associated with this gene.
- Gene
- MFGE8
- Gene Name
- milk fat globule-EGF factor 8 protein
View Full Product Details