Product Details

SNP ID
rs697228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10287787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGGGTAGGCGCTACCACTGTG[C/T]CCATTTTACAGGTGAGGACAGAGGC
Phenotype
MIM: 605353
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GHRL PubMed Links

Gene Details

Gene
GHRL
Gene Name
ghrelin/obestatin prepropeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134941.2 Intron NP_001128413.1
NM_001134944.1 Intron NP_001128416.1
NM_001134945.1 Intron NP_001128417.1
NM_001134946.1 Intron NP_001128418.1
NM_001302821.1 Intron NP_001289750.1
NM_001302822.1 Intron NP_001289751.1
NM_001302823.1 Intron NP_001289752.1
NM_001302824.1 Intron NP_001289753.1
NM_001302825.1 Intron NP_001289754.1
NM_016362.4 Intron NP_057446.1
XM_017006612.1 Intron XP_016862101.1
XM_017006613.1 Intron XP_016862102.1
Gene
GHRLOS
Gene Name
ghrelin opposite strand/antisense RNA
There are no transcripts associated with this gene.

Gene
LINC00852
Gene Name
long intergenic non-protein coding RNA 852
There are no transcripts associated with this gene.

Gene
TATDN2
Gene Name
TatD DNase domain containing 2
There are no transcripts associated with this gene.

View Full Product Details