Product Details
- SNP ID
-
rs1675510
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:98570711 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTAATATTTATAAAATATGAGGA[C/T]ACTAAGAAATATCCTATTAATATAT
- Phenotype
-
MIM: 612732
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CPOX
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111244814] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPOX
- Gene Name
- coproporphyrinogen oxidase
View Full Product Details