Product Details

SNP ID
rs812369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126481461 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGGCCTGCCCCTCTCCAATCCAC[A/C]GACATGTGGGGTGTCTGGTTTTCTG
Phenotype
MIM: 613012 MIM: 615746
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UROC1 PubMed Links

Gene Details

Gene
UROC1
Gene Name
urocanate hydratase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165974.1 2969 UTR 3 NP_001159446.1
NM_144639.2 2969 UTR 3 NP_653240.1
Gene
ZXDC
Gene Name
ZXD family zinc finger C
There are no transcripts associated with this gene.

View Full Product Details