Product Details

SNP ID
rs923
Assay Type
Validated
NCBI dbSNP Submissions
67
Location
Chr.1:111499761 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACACTGAATTAGAGAGAAAGGACAA[A/G]GGAAAAATGAAGTGGAGGAAGAGAG
Phenotype
MIM: 600445
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADORA3 PubMed Links

Gene Details

Gene
ADORA3
Gene Name
adenosine A3 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000677.3 1913 UTR 3 NP_000668.1
NM_001302678.1 1913 UTR 3 NP_001289607.1
NM_001302679.1 1913 UTR 3 NP_001289608.1
Gene
TMIGD3
Gene Name
transmembrane and immunoglobulin domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081976.2 1913 Intron NP_001075445.1
NM_001302680.1 1913 Intron NP_001289609.1
NM_020683.6 1913 Intron NP_065734.5

View Full Product Details