Product Details

SNP ID
rs1141262
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:216506089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGCTAAACTAACAAATACCTGG[A/G]AGACTTGAAAAAACAATTAAGGAAT
Phenotype
MIM: 602969
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESRRG PubMed Links

Gene Details

Gene
ESRRG
Gene Name
estrogen related receptor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134285.2 2401 UTR 3 NP_001127757.1
NM_001243505.1 2401 UTR 3 NP_001230434.1
NM_001243506.1 2401 UTR 3 NP_001230435.1
NM_001243507.1 2401 UTR 3 NP_001230436.1
NM_001243509.1 2401 UTR 3 NP_001230438.1
NM_001243510.2 2401 UTR 3 NP_001230439.1
NM_001243511.2 2401 UTR 3 NP_001230440.1
NM_001243512.1 2401 UTR 3 NP_001230441.1
NM_001243513.1 2401 UTR 3 NP_001230442.1
NM_001243514.1 2401 UTR 3 NP_001230443.1
NM_001243515.1 2401 UTR 3 NP_001230444.1
NM_001243518.1 2401 UTR 3 NP_001230447.1
NM_001243519.1 2401 UTR 3 NP_001230448.1
NM_001438.3 2401 UTR 3 NP_001429.2
NM_206594.2 2401 UTR 3 NP_996317.1
NM_206595.2 2401 UTR 3 NP_996318.1
XM_011509265.2 2401 UTR 3 XP_011507567.1
XM_011509266.2 2401 UTR 3 XP_011507568.1
XM_011509267.1 2401 UTR 3 XP_011507569.1
XM_011509268.2 2401 UTR 3 XP_011507570.1
XM_011509269.2 2401 UTR 3 XP_011507571.1
XM_011509270.1 2401 UTR 3 XP_011507572.1
XM_011509271.2 2401 UTR 3 XP_011507573.1
XM_011509274.1 2401 UTR 3 XP_011507576.1
XM_011509275.1 2401 UTR 3 XP_011507577.1
XM_011509276.1 2401 UTR 3 XP_011507578.1
XM_011509277.1 2401 UTR 3 XP_011507579.1
XM_011509278.1 2401 UTR 3 XP_011507580.1
XM_011509279.1 2401 UTR 3 XP_011507581.1
XM_011509280.2 2401 UTR 3 XP_011507582.1
XM_017000621.1 2401 UTR 3 XP_016856110.1
XM_017000622.1 2401 UTR 3 XP_016856111.1
XM_017000623.1 2401 UTR 3 XP_016856112.1
XM_017000624.1 2401 UTR 3 XP_016856113.1
XM_017000625.1 2401 UTR 3 XP_016856114.1
XM_017000626.1 2401 UTR 3 XP_016856115.1
XM_017000627.1 2401 UTR 3 XP_016856116.1
XM_017000628.1 2401 UTR 3 XP_016856117.1
XM_017000629.1 2401 UTR 3 XP_016856118.1
XM_017000630.1 2401 UTR 3 XP_016856119.1
XM_017000631.1 2401 UTR 3 XP_016856120.1
XM_017000632.1 2401 UTR 3 XP_016856121.1
XM_017000633.1 2401 UTR 3 XP_016856122.1
XM_017000634.1 2401 UTR 3 XP_016856123.1
XM_017000635.1 2401 UTR 3 XP_016856124.1
XM_017000636.1 2401 UTR 3 XP_016856125.1
XM_017000637.1 2401 UTR 3 XP_016856126.1
XM_017000638.1 2401 UTR 3 XP_016856127.1
XM_017000639.1 2401 UTR 3 XP_016856128.1
XM_017000640.1 2401 UTR 3 XP_016856129.1
XM_017000641.1 2401 UTR 3 XP_016856130.1
XM_017000642.1 2401 UTR 3 XP_016856131.1
XM_017000643.1 2401 UTR 3 XP_016856132.1
XM_017000644.1 2401 UTR 3 XP_016856133.1
XM_017000645.1 2401 UTR 3 XP_016856134.1
XM_017000646.1 2401 UTR 3 XP_016856135.1
XM_017000647.1 2401 UTR 3 XP_016856136.1
XM_017000648.1 2401 UTR 3 XP_016856137.1
XM_017000649.1 2401 UTR 3 XP_016856138.1

View Full Product Details