Product Details

SNP ID
rs1037625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32949678 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGGTGCTCATTGGACATAAAGGAG[C/T]CTTACCTTCCCCCTCAGAAAAAGTG
Phenotype
MIM: 150390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100271832 PubMed Links

Gene Details

Gene
LOC100271832
Gene Name
uncharacterized LOC100271832
There are no transcripts associated with this gene.

Gene
LTBP1
Gene Name
latent transforming growth factor beta binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000627.3 Intron NP_000618.3
NM_001166264.1 Intron NP_001159736.1
NM_001166265.1 Intron NP_001159737.1
NM_001166266.1 Intron NP_001159738.1
NM_206943.2 Intron NP_996826.2
XM_005264317.3 Intron XP_005264374.1
XM_005264318.3 Intron XP_005264375.1
XM_011532853.2 Intron XP_011531155.1
XM_011532855.2 Intron XP_011531157.1
XM_011532856.2 Intron XP_011531158.1
XM_011532857.2 Intron XP_011531159.1
XM_011532858.2 Intron XP_011531160.1
XM_011532859.2 Intron XP_011531161.1
XM_011532860.2 Intron XP_011531162.1
XM_011532861.2 Intron XP_011531163.1
XM_011532862.2 Intron XP_011531164.1
XM_017004108.1 Intron XP_016859597.1
XM_017004109.1 Intron XP_016859598.1
XM_017004110.1 Intron XP_016859599.1

View Full Product Details