Product Details
- SNP ID
-
rs1321533
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:116580022 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTATAGGTGGAGATTTTAAAATAAA[A/C]GGTAAACTACCTCATTTTTTCAGTT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RWDD1
PubMed Links
Gene Details
- Gene
- RWDD1
- Gene Name
- RWD domain containing 1
View Full Product Details