Product Details
- SNP ID
-
rs789296
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:154346951 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACAACTGAGTAACAGATTTTTAGG[A/C]TTGATATCCATCCCCCAAAATATTT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPR149
PubMed Links
Gene Details
- Gene
- GPR149
- Gene Name
- G protein-coupled receptor 149
View Full Product Details