Product Details
- SNP ID
-
rs8766
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:55228300 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCCAAGGAGAGAGGGGGAGGCTC[C/T]GAGGACACCTTCTCTTCCTCCCTTC
- Phenotype
-
MIM: 602272
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TCF4
PubMed Links
Gene Details
- Gene
- TCF4
- Gene Name
- transcription factor 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001083962.1 |
2131 |
Silent Mutation |
TCA,TCG |
S647S |
NP_001077431.1 |
NM_001243226.2 |
2131 |
Silent Mutation |
TCA,TCG |
S749S |
NP_001230155.2 |
NM_001243227.1 |
2131 |
Silent Mutation |
TCA,TCG |
S623S |
NP_001230156.1 |
NM_001243228.1 |
2131 |
Silent Mutation |
TCA,TCG |
S653S |
NP_001230157.1 |
NM_001243230.1 |
2131 |
Silent Mutation |
TCA,TCG |
S640S |
NP_001230159.1 |
NM_001243231.1 |
2131 |
Silent Mutation |
TCA,TCG |
S601S |
NP_001230160.1 |
NM_001243232.1 |
2131 |
Silent Mutation |
TCA,TCG |
S576S |
NP_001230161.1 |
NM_001243233.1 |
2131 |
Silent Mutation |
TCA,TCG |
S513S |
NP_001230162.1 |
NM_001243234.1 |
2131 |
Silent Mutation |
TCA,TCG |
S487S |
NP_001230163.1 |
NM_001243235.1 |
2131 |
Silent Mutation |
TCA,TCG |
S483S |
NP_001230164.1 |
NM_001243236.1 |
2131 |
Silent Mutation |
TCA,TCG |
S483S |
NP_001230165.1 |
NM_001306207.1 |
2131 |
Silent Mutation |
TCA,TCG |
S619S |
NP_001293136.1 |
NM_001306208.1 |
2131 |
Silent Mutation |
TCA,TCG |
S572S |
NP_001293137.1 |
NM_003199.2 |
2131 |
Silent Mutation |
TCA,TCG |
S643S |
NP_003190.1 |
XM_005266739.3 |
2131 |
Silent Mutation |
TCA,TCG |
S623S |
XP_005266796.2 |
XM_005266741.3 |
2131 |
Silent Mutation |
TCA,TCG |
S646S |
XP_005266798.1 |
XM_005266743.4 |
2131 |
Silent Mutation |
TCA,TCG |
S623S |
XP_005266800.1 |
XM_005266744.3 |
2131 |
Silent Mutation |
TCA,TCG |
S623S |
XP_005266801.1 |
XM_005266745.3 |
2131 |
Silent Mutation |
TCA,TCG |
S622S |
XP_005266802.1 |
XM_005266747.3 |
2131 |
Silent Mutation |
TCA,TCG |
S605S |
XP_005266804.1 |
XM_005266749.3 |
2131 |
Silent Mutation |
TCA,TCG |
S577S |
XP_005266806.1 |
XM_005266752.4 |
2131 |
Silent Mutation |
TCA,TCG |
S517S |
XP_005266809.1 |
XM_005266754.3 |
2131 |
Silent Mutation |
TCA,TCG |
S517S |
XP_005266811.1 |
XM_005266755.4 |
2131 |
Silent Mutation |
TCA,TCG |
S517S |
XP_005266812.1 |
XM_005266761.3 |
2131 |
Silent Mutation |
TCA,TCG |
S486S |
XP_005266818.1 |
XM_006722536.2 |
2131 |
Silent Mutation |
TCA,TCG |
S647S |
XP_006722599.1 |
XM_006722537.2 |
2131 |
Silent Mutation |
TCA,TCG |
S647S |
XP_006722600.1 |
XM_006722538.2 |
2131 |
Silent Mutation |
TCA,TCG |
S622S |
XP_006722601.1 |
XM_011526158.2 |
2131 |
Silent Mutation |
TCA,TCG |
S645S |
XP_011524460.1 |
XM_011526160.2 |
2131 |
Silent Mutation |
TCA,TCG |
S517S |
XP_011524462.1 |
XM_017025934.1 |
2131 |
Silent Mutation |
TCA,TCG |
S622S |
XP_016881423.1 |
XM_017025935.1 |
2131 |
Silent Mutation |
TCA,TCG |
S622S |
XP_016881424.1 |
XM_017025936.1 |
2131 |
Silent Mutation |
TCA,TCG |
S619S |
XP_016881425.1 |
XM_017025937.1 |
2131 |
Silent Mutation |
TCA,TCG |
S618S |
XP_016881426.1 |
XM_017025938.1 |
2131 |
Silent Mutation |
TCA,TCG |
S646S |
XP_016881427.1 |
XM_017025939.1 |
2131 |
Silent Mutation |
TCA,TCG |
S646S |
XP_016881428.1 |
XM_017025940.1 |
2131 |
Silent Mutation |
TCA,TCG |
S643S |
XP_016881429.1 |
XM_017025941.1 |
2131 |
Silent Mutation |
TCA,TCG |
S643S |
XP_016881430.1 |
XM_017025942.1 |
2131 |
Silent Mutation |
TCA,TCG |
S642S |
XP_016881431.1 |
XM_017025943.1 |
2131 |
Silent Mutation |
TCA,TCG |
S642S |
XP_016881432.1 |
XM_017025944.1 |
2131 |
Silent Mutation |
TCA,TCG |
S622S |
XP_016881433.1 |
XM_017025945.1 |
2131 |
Silent Mutation |
TCA,TCG |
S622S |
XP_016881434.1 |
XM_017025946.1 |
2131 |
Silent Mutation |
TCA,TCG |
S619S |
XP_016881435.1 |
XM_017025947.1 |
2131 |
Silent Mutation |
TCA,TCG |
S619S |
XP_016881436.1 |
XM_017025948.1 |
2131 |
Silent Mutation |
TCA,TCG |
S618S |
XP_016881437.1 |
XM_017025949.1 |
2131 |
Silent Mutation |
TCA,TCG |
S618S |
XP_016881438.1 |
XM_017025950.1 |
2131 |
Silent Mutation |
TCA,TCG |
S617S |
XP_016881439.1 |
XM_017025951.1 |
2131 |
Silent Mutation |
TCA,TCG |
S604S |
XP_016881440.1 |
XM_017025952.1 |
2131 |
Silent Mutation |
TCA,TCG |
S575S |
XP_016881441.1 |
XM_017025953.1 |
2131 |
Silent Mutation |
TCA,TCG |
S573S |
XP_016881442.1 |
XM_017025954.1 |
2131 |
Silent Mutation |
TCA,TCG |
S516S |
XP_016881443.1 |
XM_017025955.1 |
2131 |
Silent Mutation |
TCA,TCG |
S516S |
XP_016881444.1 |
XM_017025956.1 |
2131 |
Silent Mutation |
TCA,TCG |
S513S |
XP_016881445.1 |
XM_017025957.1 |
2131 |
Silent Mutation |
TCA,TCG |
S482S |
XP_016881446.1 |
View Full Product Details