Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001324191.1 | 7383 | UTR 3 | NP_001311120.1 | ||
NM_001324192.1 | 7383 | Intron | NP_001311121.1 | ||
NM_001324193.1 | 7383 | UTR 3 | NP_001311122.1 | ||
NM_024960.5 | 7383 | UTR 3 | NP_079236.3 | ||
NM_153638.3 | 7383 | UTR 3 | NP_705902.2 | ||
NM_153640.3 | 7383 | UTR 3 | NP_705904.1 | ||
XM_005260835.3 | 7383 | Intron | XP_005260892.1 | ||
XM_005260836.4 | 7383 | Intron | XP_005260893.3 | ||
XM_011529364.2 | 7383 | Intron | XP_011527666.1 | ||
XM_011529365.2 | 7383 | Intron | XP_011527667.1 | ||
XM_017028077.1 | 7383 | Intron | XP_016883566.1 | ||
XM_017028078.1 | 7383 | Intron | XP_016883567.1 | ||
XM_017028079.1 | 7383 | Intron | XP_016883568.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001134337.2 | 7383 | UTR 3 | NP_001127809.1 | ||
NM_001134338.2 | 7383 | UTR 3 | NP_001127810.1 | ||
NM_001321749.1 | 7383 | UTR 3 | NP_001308678.1 | ||
NM_007219.4 | 7383 | UTR 3 | NP_009150.1 | ||
XM_011529145.2 | 7383 | Intron | XP_011527447.1 | ||
XM_011529146.2 | 7383 | Intron | XP_011527448.1 | ||
XM_011529147.2 | 7383 | Intron | XP_011527449.1 | ||
XM_017027620.1 | 7383 | Intron | XP_016883109.1 | ||
XM_017027621.1 | 7383 | Intron | XP_016883110.1 | ||
XM_017027622.1 | 7383 | Intron | XP_016883111.1 |