Product Details

SNP ID
rs1566980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:61209783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAAATTAGGTATAAAATGAAAG[C/T]AACAATTTAAGATGTTTTAAATTAA
Phenotype
MIM: 616417
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRL3 PubMed Links

Gene Details

Gene
ADGRL3
Gene Name
adhesion G protein-coupled receptor L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322246.1 Intron NP_001309175.1
NM_001322402.1 Intron NP_001309331.1
NM_015236.5 Intron NP_056051.2
XM_011531788.2 Intron XP_011530090.1
XM_011531791.2 Intron XP_011530093.1
XM_017007929.1 Intron XP_016863418.1
XM_017007930.1 Intron XP_016863419.1
XM_017007931.1 Intron XP_016863420.1
XM_017007932.1 Intron XP_016863421.1
XM_017007933.1 Intron XP_016863422.1
XM_017007934.1 Intron XP_016863423.1
XM_017007935.1 Intron XP_016863424.1
XM_017007936.1 Intron XP_016863425.1
XM_017007937.1 Intron XP_016863426.1
XM_017007938.1 Intron XP_016863427.1
XM_017007939.1 Intron XP_016863428.1
XM_017007940.1 Intron XP_016863429.1
XM_017007941.1 Intron XP_016863430.1
XM_017007942.1 Intron XP_016863431.1

View Full Product Details