Product Details

SNP ID
rs1772159
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:205790067 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGAGAGGTGGAGGGAAGGGAAT[A/G]TGAGGAAGGGTCGTTTAGGGAAATA
Phenotype
MIM: 610801
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC41A1 PubMed Links

Gene Details

Gene
SLC41A1
Gene Name
solute carrier family 41 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173854.5 4043 UTR 3 NP_776253.3
XM_005245069.2 4043 UTR 3 XP_005245126.1

View Full Product Details