Product Details
- SNP ID
-
rs12440918
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:89696224 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTGAGAATGCTGTATAAATGGAA[C/T]TGGACTGCATATAAATTTTGAGACT
- Phenotype
-
MIM: 603866
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PEX11A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75128786] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PEX11A
- Gene Name
- peroxisomal biogenesis factor 11 alpha
There are no transcripts associated with this gene.
- Gene
- WDR93
- Gene Name
- WD repeat domain 93
View Full Product Details