Product Details

SNP ID
rs1188394
Assay Type
Functionally tested
NCBI dbSNP Submissions
74
Location
Chr.1:30711371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGTCAGTAGACCATGGCTGCCCT[A/C]GGGAGATGCTGGTTTTCATTTACAA
Phenotype
MIM: 115437
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MATN1 PubMed Links

Gene Details

Gene
MATN1
Gene Name
matrilin 1, cartilage matrix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002379.3 3736 UTR 3 NP_002370.1
Gene
MATN1-AS1
Gene Name
MATN1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details